Anti-ZBTB5 Rabbit Polyclonal Antibody

Toimittaja: United States Biological
USBI146785EA 526 EUR
USBI146785
Anti-ZBTB5 Rabbit Polyclonal Antibody
Vasta-aineet
Recommended dilution: Western Blot: 1-2?g/ml; Immunochemistry: 2.5 ?g/ml

Type: Primary
Antigen: ZBTB5 (zinc finger and BTB domain containing 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR