Anti-ASAP1 Rabbit Polyclonal Antibody

Toimittaja: United States Biological
USBI168035EA 843 EUR
USBI168035
Anti-ASAP1 Rabbit Polyclonal Antibody
Vasta-aineet
Recommended dilution: ELISA: 1:2000-1:10000; Western Blot: 1:500-1:2000

Type: Primary
Antigen: ASAP1 (ArfGAP with SH3 domain, ankyrin repeat and PH domain 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Chicken, Dog, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR