Anti-CTNNB1 Mouse monoclonal antibody [clone: 5G30]

Toimittaja: United States Biological
USBIC2096-60BEA 776 EUR
USBIC2096-60B
Anti-CTNNB1 Mouse monoclonal antibody [clone: 5G30]
Vasta-aineet
Type: Primary
Antigen: CTNNB1 (catenin (cadherin-associated protein), beta 1, 88kDa)
Clonality: Monoclonal
Clone: 5G30
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR