Anti-ELP3 Rabbit polyclonal antibody
Toimittaja: United States Biological
USBIE2249-82BEA 647 EUR
USBIE2249-82B
Anti-ELP3 Rabbit polyclonal antibody
Vasta-aineet
Recommended dilution: Western Blot: 1:500-1:3000; Immunofluorescence: 1:100-1:200; Immunochemistry: 1:100-1:200
Type: Primary
Antigen: ELP3 (elongator acetyltransferase complex subunit 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human