Anti-Elongin B Chicken polyclonal antibody

USBIE2250-05-1EA 0 EUR
USBIE2250-05-1 USBIE2250-05-200
Anti-Elongin B Chicken polyclonal antibody
Type: Primary
Antigen: Elongin B
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR