Anti-EMD Rabbit polyclonal antibody

USBIE2257-01DEA 352 EUR
USBIE2257-01D
Anti-EMD Rabbit polyclonal antibody
Recommended dilution: Western Blot: 0.5-1 μg/ml

Type: Primary
Antigen: EMD (emerin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR