Anti-EMX1 Rabbit polyclonal antibody

Toimittaja: United States Biological
USBIE2265-05EA 587 EUR
USBIE2265-05
Anti-EMX1 Rabbit polyclonal antibody
Vasta-aineet
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500

Type: Primary
Antigen: EMX1 (empty spiracles homeobox 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR