Anti-EMX1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Toimittaja: United States Biological
USBIE2265-05-APEA 852 EUR
USBIE2265-05-AP
Anti-EMX1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Vasta-aineet
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500
Do not freeze!
Type: Primary
Antigen: EMX1 (empty spiracles homeobox 1)
Clonality: polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human