Anti-EN2, CT-2 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Toimittaja: United States Biological
USBIE2270-10C-PEEA 852 EUR
USBIE2270-10C-PE
Anti-EN2, CT-2 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Vasta-aineet
Recommended dilution: FLISA: 1:1000; Western Blot: 1:50-1:100
Do not freeze!
Type: Primary
Antigen: EN2, CT-2
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human