Anti-EN2 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Toimittaja: United States Biological
USBIE2270-10E-FITCEA 852 EUR
USBIE2270-10E-FITC
Anti-EN2 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Vasta-aineet
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500
Do not freeze!
Type: Primary
Antigen: EN2 (engrailed homeobox 2)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse