Anti-GSK3A Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Toimittaja: United States Biological
USBIG8170-42N-HRPEA 852 EUR
USBIG8170-42N-HRP
Anti-GSK3A Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Vasta-aineet
Recommended dilution: ELISA: 1:1000; Immunochemistry: 1:50-1:100

Type: Primary
Antigen: GSK3A (glycogen synthase kinase 3 alpha)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR