Anti-Protein Tyrosine Phosphatase Type 4A, Member 2 Chicken Polyclonal Antibody
Toimittaja: United States Biological
USBIP9109-46EA 729 EUR
USBIP9109-46
Anti-Protein Tyrosine Phosphatase Type 4A, Member 2 Chicken Polyclonal Antibody
Vasta-aineet
Recommended dilution: Western Blot: 1:1000-1:2000
Type: Primary
Antigen: Protein Tyrosine Phosphatase Type 4A, Member 2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse, Rat