Anti-PTP4A2 Rabbit Polyclonal Antibody

Toimittaja: United States Biological
USBIP9109-46AEA 587 EUR
USBIP9109-46A
Anti-PTP4A2 Rabbit Polyclonal Antibody
Vasta-aineet
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-1:100

Type: Primary
Antigen: PTP4A2 (protein tyrosine phosphatase type IVA, member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR