Anti-PRL3 Mouse Monoclonal Antibody [clone: 10i66]
Toimittaja: United States Biological
USBIP9109-48C1EA 542 EUR
USBIP9109-48C1
Anti-PRL3 Mouse Monoclonal Antibody [clone: 10i66]
Vasta-aineet
  • Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500
Type: Primary
Antigen: PRL3
Clonality: monoclonal
Clone: 10i66
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse