Anti-PRL3 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Toimittaja: United States Biological
USBIP9109-48C2FITCEA 852 EUR
USBIP9109-48C2FITC
Anti-PRL3 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Vasta-aineet
Type: Primary
Antigen: PRL3
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human