Anti-PRL3 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Toimittaja: United States Biological
USBIP9109-48C2-HRPEA 852 EUR
USBIP9109-48C2-HRP
Anti-PRL3 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Vasta-aineet
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500
Type: Primary
Antigen: PRL3
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human