Anti-PRL3 Rabbit Polyclonal Antibody

Toimittaja: United States Biological
USBIP9109-48C3EA 587 EUR
USBIP9109-48C3
Anti-PRL3 Rabbit Polyclonal Antibody
Vasta-aineet
Recommended dilution: Western Blot: 1:1000

Type: Primary
Antigen: PRL3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR