Anti-Prothrombin Sheep Polyclonal Antibody (FITC (Fluorescein))
Toimittaja: United States Biological
USBIP9115-08DEA 837 EUR
USBIP9115-08D
Anti-Prothrombin Sheep Polyclonal Antibody (FITC (Fluorescein))
Vasta-aineet
Type: Primary
Antigen: Prothrombin
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Sheep
Isotype:
Reactivity: Mouse