Anti-Prothrombin Sheep Polyclonal Antibody
Toimittaja: United States Biological
USBIP9115-08EEA 755 EUR
USBIP9115-08E
Anti-Prothrombin Sheep Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: Prothrombin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity: Mouse