Anti-PCDHA2 Mouse Polyclonal Antibody

Toimittaja: United States Biological
USBIP9119-11EA 555 EUR
USBIP9119-11
Anti-PCDHA2 Mouse Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: PCDHA2 (protocadherin alpha 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR