Anti-PTMA Goat Polyclonal Antibody

Toimittaja: United States Biological
USBIP9119-35CEA 551 EUR
USBIP9119-35C
Anti-PTMA Goat Polyclonal Antibody
Vasta-aineet
Recommended dilution: ELISA: 1:1000

Type: Primary
Antigen: PTMA (prothymosin, alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Dog, Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR