Anti-SNRPD2 Rabbit Polyclonal Antibody

Toimittaja: United States Biological
USBIS1014-96SEA 587 EUR
USBIS1014-96S
Anti-SNRPD2 Rabbit Polyclonal Antibody
Vasta-aineet
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-1:100

Type: Primary
Antigen: SNRPD2 (small nuclear ribonucleoprotein D2 polypeptide 16.5kDa)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR