Anti-SNX16 Goat Polyclonal Antibody

Toimittaja: United States Biological
USBIS1014-97M1EA 551 EUR
USBIS1014-97M1
Anti-SNX16 Goat Polyclonal Antibody
Vasta-aineet
Recommended dilution: ELISA: 1: 64000; Western Blot: 0. 05-0.2?g/ml

Type: Primary
Antigen: SNX16 (sorting nexin 16)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR