Anti-SNX16 Goat Polyclonal Antibody

Toimittaja: United States Biological
USBIS1014-97M3EA 633 EUR
USBIS1014-97M3
Anti-SNX16 Goat Polyclonal Antibody
Vasta-aineet
Recommended dilution: ELISA: 1:64000; Western Blot: 1:64000

Type: Primary
Antigen: SNX16 (sorting nexin 16)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR