Anti-TRPC3 Rabbit polyclonal antibody
USBIT8666-14DEA 352 EUR
USBIT8666-14D
Anti-TRPC3 Rabbit polyclonal antibody
Recommended dilution: Western Blot: 0.5-1 μg/ml; Immunochemistry: 2μg/ml
Type: Primary
Antigen: TRPC3 (transient receptor potential cation channel, subfamily C, member 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat