Anti-TRPC6 Rabbit Polyclonal Antibody

Toimittaja: United States Biological
USBIT8666-30AEA 754 EUR
USBIT8666-30A
Anti-TRPC6 Rabbit Polyclonal Antibody
Vasta-aineet
Recommended dilution: Immunochemistry: 5 ?g/ml; Western Blot: 1:500-1:2000

Type: Primary
Antigen: TRPC6 (transient receptor potential cation channel, subfamily C, member 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR