Anti-TRPC6 Rabbit polyclonal antibody
USBIT8666-30FEA 514 EUR
USBIT8666-30F
Anti-TRPC6 Rabbit polyclonal antibody
Recommended dilution: Western Blot: 0.5-1 μg/ml
Type: Primary
Antigen: TRPC6 (transient receptor potential cation channel, subfamily C, member 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse