Anti-TRPC6 Goat Polyclonal Antibody

Toimittaja: United States Biological
USBIT8666-30LEA 551 EUR
USBIT8666-30L
Anti-TRPC6 Goat Polyclonal Antibody
Vasta-aineet
Recommended dilution: ELISA: 1:2000

Type: Primary
Antigen: TRPC6 (transient receptor potential cation channel, subfamily C, member 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Dog, Human, Mouse, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR