Anti-TRPM2 Goat Polyclonal Antibody

Toimittaja: United States Biological
USBIT8666-35FEA 633 EUR
USBIT8666-35F
Anti-TRPM2 Goat Polyclonal Antibody
Vasta-aineet
Recommended dilution: Immunochemistry: 3. 75 ?g/ml

Type: Primary
Antigen: TRPM2 (transient receptor potential cation channel, subfamily M, member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR