Anti-TRPM5 Goat polyclonal antibody

USBIT8666-37MEA 401 EUR
USBIT8666-37M
Anti-TRPM5 Goat polyclonal antibody
Recommended dilution: ELISA: 1:128000; Western Blot: 1-3μg/ml

Type: Primary
Antigen: TRPM5 (transient receptor potential cation channel, subfamily M, member 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR