Anti-TRPM6 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Toimittaja: United States Biological
USBIT8666-37Q-APEA 852 EUR
USBIT8666-37Q-AP
Anti-TRPM6 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Vasta-aineet
Recommended dilution: ELISA: 1:1000; Immunochemistry: 1:50-1:100
Do not freeze!
Type: Primary
Antigen: TRPM6 (transient receptor potential cation channel, subfamily M, member 6)
Clonality: polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human