Anti-TRPM6 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Toimittaja: United States Biological
USBIT8666-37Q-APCEA 852 EUR
USBIT8666-37Q-APC
Anti-TRPM6 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Vasta-aineet
Recommended dilution: FLISA: 1:1000; Immunochemistry: 1:50-1:100
Do not freeze!
Type: Primary
Antigen: TRPM6 (transient receptor potential cation channel, subfamily M, member 6)
Clonality: polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human