Anti-TRPM7 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Toimittaja: United States Biological
USBIT8666-37V-HRPEA 852 EUR
USBIT8666-37V-HRP
Anti-TRPM7 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Vasta-aineet
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-1:200; Immunochemistry: 1:50-1:100

Type: Primary
Antigen: TRPM7 (transient receptor potential cation channel, subfamily M, member 7)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR