Anti-TRPM8 Rabbit Polyclonal Antibody (Biotin)
Toimittaja: United States Biological
USBIT8666-38LEA 1090 EUR
USBIT8666-38L
Anti-TRPM8 Rabbit Polyclonal Antibody (Biotin)
Vasta-aineet
Type: Primary
Antigen: TRPM8 (transient receptor potential cation channel, subfamily M, member 8)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat