Anti-TRRAP Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Toimittaja: United States Biological
USBIT8666-40A-APEA 616 EUR
USBIT8666-40A-AP
Anti-TRRAP Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Vasta-aineet
Recommended dilution: ELISA: 1:1000; Immunochemistry: 1:50-1:100
Do not freeze!
Type: Primary
Antigen: TRRAP (transformation/transcription domain-associated protein)
Clonality: polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human