Anti-TRRAP Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Toimittaja: United States Biological
USBIT8666-40A-APCEA 616 EUR
USBIT8666-40A-APC
Anti-TRRAP Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Vasta-aineet
Recommended dilution: FLISA: 1:1000; Immunochemistry: 1:50-1:100
Do not freeze!

Type: Primary
Antigen: TRRAP (transformation/transcription domain-associated protein)
Clonality: polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR