Anti-UTS2 Rabbit Polyclonal Antibody

Toimittaja: United States Biological
USBIU2800-02CEA 979 EUR
USBIU2800-02C
Anti-UTS2 Rabbit Polyclonal Antibody
Vasta-aineet
Recommended dilution: Radio Immunoassay: 0.24?g/ml

Type: Primary
Antigen: UTS2 (urotensin 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR