Culture media in sachets
Toimittaja: Scharlab
SCAR01-111BA05EA 193 EUR
SCAR01-144BA05 SCAR02-494BA05 SCAR01-265BA05 SCAR02-200BA05 SCAR01-540BA05 SCAR03-156BA05 SCAR01-111BA05 SCAR01-165BA05 SCAR064-BA04-2 SCAR01-200BA05 SCAR01-412BA05 SCAR01-161BA05 SCAR01-589BA05 SCAR02-277BA05 SCAR02-599BA05 SCAR01-034BA05 SCAR01-309BA05 SCAR01-030BA05 SCAR02-060BA05
Culture media in sachets
Elatusaineet mikrobiologiaan Valmisteltu viljelyaine
Dehydrated media in sachets are an ideal solution for media used in smaller quantities.
  • Fast
  • Simple
  • Safe
  • Less waste