Muffel tray
Toimittaja: TQS-Quartz
TSLA71331EA 624 EUR
TSLA71331
Muffel tray
Haihdutusmaljat/Tarjottimet/Altaat Laboratorioalustat/-astiat
Muffel trays made of quartz, translucent.
  • Excellent chemical resistance
  • Excellent temperature resistance
  • With 4 holes