Anti-FSHB Mouse Monoclonal Antibody [clone: 4E8]

Toimittaja: OriGene
ORIGCF501624EA 770 EUR
ORIGCF501624
Anti-FSHB Mouse Monoclonal Antibody [clone: 4E8]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: FSHB
Clonality: Monoclonal
Clone: 4E8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR