Anti-NSMCE2 Mouse Monoclonal Antibody [clone: 4A2]

Toimittaja: OriGene
ORIGCF501632EA 749 EUR
ORIGCF501632
Anti-NSMCE2 Mouse Monoclonal Antibody [clone: 4A2]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: NSMCE2
Clonality: Monoclonal
Clone: 4A2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR