Anti-FGF21 Mouse Monoclonal Antibody [clone: 1B4]

Toimittaja: OriGene
ORIGCF502787EA 770 EUR
ORIGCF502787
Anti-FGF21 Mouse Monoclonal Antibody [clone: 1B4]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: FGF21
Clonality: Monoclonal
Clone: 1B4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR