Anti-UBOX5 Mouse Monoclonal Antibody [clone: 7G1]

Toimittaja: OriGene
ORIGCF502810EA 749 EUR
ORIGCF502810
Anti-UBOX5 Mouse Monoclonal Antibody [clone: 7G1]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: UBOX5
Clonality: Monoclonal
Clone: 7G1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR