Anti-FRK Mouse Monoclonal Antibody [clone: 3E2]

Toimittaja: OriGene
ORIGCF501052EA 749 EUR
ORIGCF501052
Anti-FRK Mouse Monoclonal Antibody [clone: 3E2]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: FRK
Clonality: Monoclonal
Clone: 3E2
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR