Anti-PIM2 Mouse Monoclonal Antibody [clone: 5B5]

Toimittaja: OriGene
ORIGCF501061EA 770 EUR
ORIGCF501061
Anti-PIM2 Mouse Monoclonal Antibody [clone: 5B5]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100; Immunoprecipitation: 2?g/500?l

Type: Primary
Antigen: PIM2
Clonality: Monoclonal
Clone: 5B5
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR