Anti-MTRF1L Mouse Monoclonal Antibody [clone: 2C6]

Toimittaja: OriGene
ORIGCF501066EA 749 EUR
ORIGCF501066
Anti-MTRF1L Mouse Monoclonal Antibody [clone: 2C6]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100 Flow Cytometry: 1:100

Type: Primary
Antigen: MTRF1L
Clonality: Monoclonal
Clone: 2C6
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR