Anti-TPMT Mouse Monoclonal Antibody [clone: 4B1]
Toimittaja: OriGene
ORIGCF501069EA 749 EUR
ORIGCF501069
Anti-TPMT Mouse Monoclonal Antibody [clone: 4B1]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50
Type: Primary
Antigen: TPMT
Clonality: Monoclonal
Clone: 4B1
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human