Anti-HP Mouse Monoclonal Antibody [clone: 1B1]

Toimittaja: OriGene
ORIGCF501700EA 749 EUR
ORIGCF501700
Anti-HP Mouse Monoclonal Antibody [clone: 1B1]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:200-1000; Flow Cytometry: 1:100

Type: Primary
Antigen: HP
Clonality: Monoclonal
Clone: 1B1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR