Anti-PUS7 Mouse Monoclonal Antibody [clone: 4A2]

Toimittaja: OriGene
ORIGCF502049EA 749 EUR
ORIGCF502049
Anti-PUS7 Mouse Monoclonal Antibody [clone: 4A2]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-2000

Type: Primary
Antigen: PUS7
Clonality: Monoclonal
Clone: 4A2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR