Anti-CALCOCO2 Mouse Monoclonal Antibody [clone: 3F2]

Toimittaja: OriGene
ORIGCF502054EA 770 EUR
ORIGCF502054
Anti-CALCOCO2 Mouse Monoclonal Antibody [clone: 3F2]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: CALCOCO2
Clonality: Monoclonal
Clone: 3F2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR